4-Formyl-2-methylbenzonitrile ≥97%

Supplier: AOBChem USA

Warning

46106-1G 46106-25G 46106-5G
76932-578EA 70.02 USD
76932-578 76932-580 76932-582
4-Formyl-2-methylbenzonitrile ≥97%
4-Formyl-2-methylbenzonitrile

Precaución: for Research Use Only

Formula: C₉H₇NO
MW: 145.158 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18394298
CAS Number: 27613-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR