5-Bromo-[1,1'-biphenyl]-2-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

45994-1G 45994-500MG 45994-5G
76932-564EA 660.95 USD
76932-564 76932-566 76932-568
5-Bromo-[1,1'-biphenyl]-2-carboxylic acid ≥97%
5-Bromo-[1,1'-biphenyl]-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₉BrO₂
MW: 277.113 g/mol
Storage Temperature: Ambient
CAS Number: 99514-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR