5-Bromo-2,4-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

45845-1G 45845-25G 45845-500MG 45845-5G
76932-548EA 332.07 USD
76932-548 76932-550 76932-552 76932-554
5-Bromo-2,4-dimethylbenzoic acid ≥97%
5-Bromo-2,4-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD24107212
CAS Number: 842136-27-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR