5-Bromo-2-(cyclopropylamino)pyrimidine ≥97%

Supplier: AOBChem USA

Warning

45671-1G 45671-25G 45671-5G
76932-512EA 88.06 USD
76932-512 76932-514 76932-516
5-Bromo-2-(cyclopropylamino)pyrimidine ≥97%
5-Bromo-2-(cyclopropylamino)pyrimidine

Precaución: for Research Use Only

Formula: C₇H₈BrN₃
Storage Temperature: Refrigerator
MDL Number: MFCD03646032
CAS Number: 886366-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR