5-Bromo-1-fluoro-2-isopropoxy-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

45645-1G 45645-25G 45645-500MG 45645-5G
76932-490EA 151.71 USD
76932-490 76932-492 76932-494 76932-496
5-Bromo-1-fluoro-2-isopropoxy-3-(trifluoromethyl)benzene ≥97%
5-Bromo-1-fluoro-2-isopropoxy-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₉BrF₄O
MW: 301.075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707136
CAS Number: 2413441-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR