4-Bromo-5-fluoro-2-methylphenol ≥97%

Supplier: AOBChem USA

Warning

45368-1G 45368-25G 45368-500MG 45368-5G
76932-474EA 346.92 USD
76932-474 76932-476 76932-478 76932-480
4-Bromo-5-fluoro-2-methylphenol ≥97%
4-Bromo-5-fluoro-2-methylphenol

Precaución: for Research Use Only

Formula: C₇H₆BrFO
MW: 205.024 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22414503
CAS Number: 1600166-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR