2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Formyl-3-methoxyphenylboronic acid pinacol ester

44792-1G 44792-25G 44792-500MG 44792-5G
76932-458EA 228.1 USD
76932-458 76932-460 76932-462 76932-464
2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥97%
2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Formula: C₁₄H₁₉BO₄
MW: 262.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18732927
CAS Number: 956431-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR