4-Chloro-5-fluoro-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

44571-1G 44571-500MG 44571-5G
76932-446EA 676.86 USD
76932-446 76932-448 76932-450
4-Chloro-5-fluoro-2-methylphenylboronic acid ≥97%
4-Chloro-5-fluoro-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BClFO₂
Storage Temperature: Ambient
MDL Number: MFCD23712926
CAS Number: 2055778-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR