5-Bromo-4-methylthiophene-2-boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (5-Bromo-4-methylthiophen-2-yl)boronic acid

41420-1G 41420-25G 41420-500MG 41420-5G
76932-408EA 125.19 USD
76932-408 76932-410 76932-412 76932-414
5-Bromo-4-methylthiophene-2-boronic acid ≥97%
5-Bromo-4-methylthiophene-2-boronic acid

Precaución: for Research Use Only

Formula: C₅H₆BBrO₂S
MW: 220.88 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12913941
CAS Number: 1217501-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR