2-(4-Bromo-2-fluoro-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-fluoro-5-(trifluoromethyl)phenylboronic acid, pinacol ester

40386-1G 40386-25G 40386-500MG 40386-5G
76932-396EA 245.07 USD
76932-396 76932-398 76932-400 76932-402
2-(4-Bromo-2-fluoro-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-Bromo-2-fluoro-5-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₃H₁₄BBrF₄O₂
MW: 368.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20441906
CAS Number: 2096337-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR