4-Fluoro-2-isopropoxy-5-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

40261-1G 40261-25G 40261-5G
76932-382EA 66.84 USD
76932-382 76932-384 76932-386
4-Fluoro-2-isopropoxy-5-methylphenylboronic acid ≥97%
4-Fluoro-2-isopropoxy-5-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD20231447
CAS Number: 2096336-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR