4-Fluoro-2-methoxy-5-methylphenylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

40230-1G 40230-25G 40230-500MG 40230-5G
76932-366EA 292.81 USD
76932-366 76932-368 76932-370 76932-372
4-Fluoro-2-methoxy-5-methylphenylboronic acid, pinacol ester ≥97%
4-Fluoro-2-methoxy-5-methylphenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₀BFO₃
MW: 266.116 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19981541
CAS Number: 2096334-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR