5-(2-Methoxyethoxy)pyridine-3-boronic acid ≥97%

Supplier: AOBChem USA

Warning

39872-1G 39872-25G 39872-500MG 39872-5G
76932-338EA 423.3 USD
76932-338 76932-340 76932-342 76932-344
5-(2-Methoxyethoxy)pyridine-3-boronic acid ≥97%
5-(2-Methoxyethoxy)pyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₂BNO₄
Storage Temperature: Refrigerator
MDL Number: MFCD13183780
CAS Number: 1015229-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR