5-Bromo-6-methoxypyridine-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-6-methoxynicotinic acid

38777-25G 38777-5G
76932-300EA 410.57 USD
76932-300 76932-302
5-Bromo-6-methoxypyridine-3-carboxylic acid ≥97%
5-Bromo-6-methoxypyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD11226508
CAS Number: 1186194-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR