5-Bromo-1-ethylpyridin-2(1H)-one ≥97% research grade

Supplier: AOBChem USA

Warning

38739-25G 38739-5G
76932-290EA 318.27 USD
76932-290 76932-292
5-Bromo-1-ethylpyridin-2(1H)-one ≥97% research grade
5-Bromo-1-ethylpyridin-2(1H)-one
Formula: C₇H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD00456284
CAS Number: 63785-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR