4-Butoxy-2,3-difluorophenol ≥95%

Supplier: AOBChem USA

Warning

38545-1G 38545-25G 38545-500MG 38545-5G
76932-250EA 233.4 USD
76932-250 76932-252 76932-254 76932-256
4-Butoxy-2,3-difluorophenol ≥95%
4-Butoxy-2,3-difluorophenol
Formula: C₁₀H₁₂F₂O₂
MW: 202.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368023
CAS Number: 136239-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR