5-Chloro-N-cyclopropyl-2-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

38490-1G 38490-25G 38490-5G
76932-236EA 106.09 USD
76932-236 76932-238 76932-240
5-Chloro-N-cyclopropyl-2-(trifluoromethyl)benzamide ≥97%
5-Chloro-N-cyclopropyl-2-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₁H₉ClF₃NO
Storage Temperature: Refrigerator
MDL Number: MFCD32707088
CAS Number: 2413441-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR