4-Chloro-2,5-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

38402-1G 38402-25G 38402-500MG 38402-5G
76932-220EA 381.93 USD
76932-220 76932-222 76932-224 76932-226
4-Chloro-2,5-dimethylbenzaldehyde ≥97%
4-Chloro-2,5-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO
MW: 168.62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11845878
CAS Number: 866490-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR