4-Butoxy-3,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

38205-1G 38205-25G 38205-5G
76932-194EA 84.88 USD
76932-194 76932-196 76932-198
4-Butoxy-3,5-dimethylbenzoic acid ≥97%
4-Butoxy-3,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
Storage Temperature: Refrigerator
MDL Number: MFCD11188419
CAS Number: 1038731-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR