4-Fluoro-2,3-dimethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

38129-1G 38129-500MG 38129-5G
76932-182EA 558.04 USD
76932-182 76932-184 76932-186
4-Fluoro-2,3-dimethoxybenzaldehyde ≥97%
4-Fluoro-2,3-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.164 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22417610
CAS Number: 1364687-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR