4-Bromo-2-fluoro-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

38098-1G 38098-25G 38098-500MG 38098-5G
76932-168EA 144.29 USD
76932-168 76932-170 76932-172 76932-174
4-Bromo-2-fluoro-3-methylbenzaldehyde ≥97%
4-Bromo-2-fluoro-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO
MW: 217.035 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28348517
CAS Number: 1784897-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR