5-Bromo-4-ethoxy-2,3-difluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37916-1G 37916-25G 37916-500MG 37916-5G
76932-142EA 280.08 USD
76932-142 76932-144 76932-146 76932-148
5-Bromo-4-ethoxy-2,3-difluorobenzoic acid ≥97%
5-Bromo-4-ethoxy-2,3-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇BrF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD32632278
CAS Number: 2404734-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR