4'-Formyl-[1,1'-biphenyl]-2-carbonitrile ≥95%

Supplier: AOBChem USA

Warning

37774-1G 37774-25G 37774-500MG 37774-5G
76932-112EA 263.11 USD
76932-112 76932-114 76932-116 76932-118
4'-Formyl-[1,1'-biphenyl]-2-carbonitrile ≥95%
4'-Formyl-[1,1'-biphenyl]-2-carbonitrile
Formula: C₁₄H₉NO
Storage Temperature: Refrigerator
MDL Number: MFCD05979418
CAS Number: 135689-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR