4-Butoxy-2,6-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37681-1G 37681-25G 37681-500MG 37681-5G
76932-084EA 209 USD
76932-084 76932-086 76932-088 76932-090
4-Butoxy-2,6-dimethylbenzoic acid ≥97%
4-Butoxy-2,6-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MW: 222.28 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32632255
CAS Number: 92156-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR