5-Chloro-2-propoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37536-1G 37536-25G 37536-5G
76932-076EA 117.76 USD
76932-076 76932-078 76932-080
5-Chloro-2-propoxybenzoic acid ≥97%
5-Chloro-2-propoxybenzoic acid
Formula: C₁₀H₁₁ClO₃
Storage Temperature: Ambient
MDL Number: MFCD02729207
CAS Number: 62176-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR