4-Ethoxy-2,3-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

37462-25G 37462-5G
76932-056EA 590.93 USD
76932-056 76932-058
4-Ethoxy-2,3-difluorobenzaldehyde ≥97%
4-Ethoxy-2,3-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MW: 186.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258700
CAS Number: 126162-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR