5-Bromo-2-chloro-4-(methoxymethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

37426-1G 37426-500MG 37426-5G
76932-038EA 510.3 USD
76932-038 76932-040 76932-042
5-Bromo-2-chloro-4-(methoxymethyl)pyridine ≥97%
5-Bromo-2-chloro-4-(methoxymethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₇BrClNO
MW: 236.494 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32632216
CAS Number: 2356252-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR