5-(4-Bromophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

37404-1G 37404-500MG 37404-5G
76932-026EA 471.04 USD
76932-026 76932-028 76932-030
5-(4-Bromophenyl)thiazol-2-amine ≥97%
5-(4-Bromophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₇BrN₂S
MW: 255.134 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02690769
CAS Number: 73040-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR