5-Ethyl-2-hydroxybenzenecarbaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 5-Ethyl-2-hydroxybenzaldehyde

37319-1G 37319-25G 37319-500MG 37319-5G
76932-014EA 138.98 USD
76932-014 76932-016 76932-018 76932-020
5-Ethyl-2-hydroxybenzenecarbaldehyde ≥97% research grade
5-Ethyl-2-hydroxybenzenecarbaldehyde
Formula: C₉H₁₀O₂
Storage Temperature: Refrigerator
MDL Number: MFCD06739514
CAS Number: 52411-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR