4-Bromo-2-fluoro-3-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37221-1G 37221-25G 37221-500MG 37221-5G
76931-984EA 142.17 USD
76931-984 76931-986 76931-988 76931-990
4-Bromo-2-fluoro-3-methoxybenzoic acid ≥97%
4-Bromo-2-fluoro-3-methoxybenzoic acid
Formula: C₈H₆BrFO₃
MW: 249.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06656802
CAS Number: 194804-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR