3-Amino-5-bromo-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-1H-indazol-3-amine

37070-25G 37070-5G
76931-972EA 204.76 USD
76931-972 76931-974
3-Amino-5-bromo-1H-indazole ≥97%
3-Amino-5-bromo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₆BrN₃
MW: 212.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03426696
CAS Number: 61272-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR