4-Chloro-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

36896-25G 36896-5G
76931-946EA 375.56 USD
76931-946 76931-948
4-Chloro-2-methoxybenzaldehyde ≥97%
4-Chloro-2-methoxybenzaldehyde
Formula: C₈H₇ClO₂
MW: 170.5900 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08703355
CAS Number: 53581-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR