4-Chloro-4'-methoxy-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

36787-1G 36787-25G 36787-500MG 36787-5G
76931-922EA 221.73 USD
76931-922 76931-924 76931-926 76931-928
4-Chloro-4'-methoxy-1,1'-biphenyl ≥95%
4-Chloro-4'-methoxy-1,1'-biphenyl
Formula: C₁₃H₁₁ClO
MW: 218.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318447
CAS Number: 58970-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR