4'-Methoxy-[1,1'-biphenyl]-4-carbonitrile ≥97%

Supplier: AOBChem USA

Warning

36773-1G 36773-25G 36773-5G
76931-894EA 96.55 USD
76931-894 76931-896 76931-898
4'-Methoxy-[1,1'-biphenyl]-4-carbonitrile ≥97%
4'-Methoxy-[1,1'-biphenyl]-4-carbonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO
MW: 209.243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01003080
CAS Number: 58743-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR