4-Bromo-N-cyclopropyl-2-fluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

36706-25G 36706-5G
76931-882EA 573.95 USD
76931-882 76931-884
4-Bromo-N-cyclopropyl-2-fluorobenzamide ≥97%
4-Bromo-N-cyclopropyl-2-fluorobenzamide
Formula: C₁₀H₉BrFNO
Storage Temperature: Ambient
MDL Number: MFCD09943270
CAS Number: 877383-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR