4'-Methylbiphenyl-4-carboxaldehyde ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 4'-Methyl-[1,1'-biphenyl]-4-carbaldehyde

36547-25G 36547-5G
76931-866EA 418 USD
76931-866 76931-868
4'-Methylbiphenyl-4-carboxaldehyde ≥95%
4'-Methylbiphenyl-4-carboxaldehyde
Formula: C₁₄H₁₂O
MW: 196.249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02258938
CAS Number: 36393-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR