4-Bromo-N,N-dimethyl-2-fluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-fluoro-N,N-dimethylbenzamide

36527-25G 36527-5G
76931-858EA 559.1 USD
76931-858 76931-860
4-Bromo-N,N-dimethyl-2-fluorobenzamide ≥97%
4-Bromo-N,N-dimethyl-2-fluorobenzamide

Precaución: for Research Use Only

Formula: C₉H₉BrFNO
MW: 246.0763 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878360
CAS Number: 749927-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR