5-Bromo-2,3,4-trifluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

36501-1G 36501-25G 36501-500MG 36501-5G
76931-848EA 118.83 USD
76931-848 76931-850 76931-852 76931-854
5-Bromo-2,3,4-trifluorobenzaldehyde ≥95%
5-Bromo-2,3,4-trifluorobenzaldehyde
Formula: C₇H₂BrF₃O
MW: 238.989 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12756767
CAS Number: 914935-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR