5-Bromo-1-(difluoromethoxy)-2-fluoro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

36423-1G 36423-25G 36423-500MG 36423-5G
76931-838EA 220.67 USD
76931-838 76931-840 76931-842 76931-844
5-Bromo-1-(difluoromethoxy)-2-fluoro-3-methylbenzene ≥97%
5-Bromo-1-(difluoromethoxy)-2-fluoro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₆BrF₃O
MW: 255.032 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32632011
CAS Number: 2167653-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR