5-Bromo-2-(1,3-oxazol-5-yl)pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-(5-Bromopyridin-2-yl)oxazole

36096-1G 36096-25G 36096-500MG 36096-5G
76931-794EA 190.97 USD
76931-794 76931-796 76931-798 76931-800
5-Bromo-2-(1,3-oxazol-5-yl)pyridine ≥97%
5-Bromo-2-(1,3-oxazol-5-yl)pyridine

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD27950166
CAS Number: 380380-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR