5-(Difluoromethoxy)-2-fluoro-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

35917-1G 35917-500MG 35917-5G
76931-780EA 718.23 USD
76931-780 76931-782 76931-784
5-(Difluoromethoxy)-2-fluoro-4-iodopyridine ≥97%
5-(Difluoromethoxy)-2-fluoro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₆H₃F₃INO
MW: 288.994 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28789545
CAS Number: 1806334-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR