5-[4-(Trifluoromethyl)phenyl]-1,3-oxazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-(4-(Trifluoromethyl)phenyl)oxazole

35826-1G 35826-25G 35826-500MG 35826-5G
76931-764EA 183.54 USD
76931-764 76931-766 76931-768 76931-770
5-[4-(Trifluoromethyl)phenyl]-1,3-oxazole ≥97%
5-[4-(Trifluoromethyl)phenyl]-1,3-oxazole

Precaución: for Research Use Only

Formula: C₁₀H₆F₃NO
Storage Temperature: Refrigerator
MDL Number: MFCD13176792
CAS Number: 87150-14-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR