5-(2-Methoxyphenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35822-1G 35822-25G 35822-500MG 35822-5G
76931-748EA 165.51 USD
76931-748 76931-750 76931-752 76931-754
5-(2-Methoxyphenyl)oxazole ≥97%
5-(2-Methoxyphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
MW: 175.184 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09749680
CAS Number: 391927-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR