5-(3-Methoxyphenyl)-1,3-oxazole ≥95%

Supplier: AOBChem USA

Warning

35722-1G 35722-25G 35722-500MG 35722-5G
76931-700EA 201.58 USD
76931-700 76931-702 76931-704 76931-706
5-(3-Methoxyphenyl)-1,3-oxazole ≥95%

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09749681
CAS Number: 848608-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR