5-Bromo-2-fluoro-3-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

35403-1G 35403-25G 35403-500MG 35403-5G
76931-660EA 168.69 USD
76931-660 76931-662 76931-664 76931-666
5-Bromo-2-fluoro-3-methoxyphenol ≥97%
5-Bromo-2-fluoro-3-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆BrFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD28738849
CAS Number: 1782384-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR