2-(4-(Methoxymethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Methoxymethoxyphenylboronic acid, pinacol ester

35378-1G 35378-25G 35378-500MG 35378-5G
76931-646EA 129.43 USD
76931-646 76931-648 76931-650 76931-652
2-(4-(Methoxymethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(4-(Methoxymethoxy)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₄H₂₁BO₄
MW: 264.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08690234
CAS Number: 936250-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR