4-Bromo-3-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

35352-1G 35352-25G 35352-500MG 35352-5G
76931-634EA 379.81 USD
76931-634 76931-636 76931-638 76931-640
4-Bromo-3-formylbenzoic acid ≥97%
4-Bromo-3-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅BrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD20485838
CAS Number: 776315-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR