5-Bromo-8-fluoroquinoline ≥97%

Supplier: AOBChem USA

Warning

35296-1G 35296-25G 35296-5G
76931-620EA 84.88 USD
76931-620 76931-622 76931-624
5-Bromo-8-fluoroquinoline ≥97%
5-Bromo-8-fluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrFN
MW: 226.0500 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11855895
CAS Number: 1133115-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR