4-Ethoxy-3,5-dimethylphenol ≥97%

Supplier: AOBChem USA

Warning

35218-1G 35218-25G 35218-500MG 35218-5G
76931-604EA 287.51 USD
76931-604 76931-606 76931-608 76931-610
4-Ethoxy-3,5-dimethylphenol ≥97%
4-Ethoxy-3,5-dimethylphenol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
MW: 166.217 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24726029
CAS Number: 217448-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR