4-Bromo-3-fluoro-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

35106-1G 35106-25G 35106-500MG 35106-5G
76931-588EA 183.54 USD
76931-588 76931-590 76931-592 76931-594
4-Bromo-3-fluoro-2-methylbenzaldehyde ≥97%
4-Bromo-3-fluoro-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO
Storage Temperature: Refrigerator
MDL Number: MFCD28739695
CAS Number: 1807174-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR